Product Details

SNP ID
rs12653537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150778602 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGCTACGCGGCGAGTGTGTGGCAA[C/G]GTGGAAGGGGAGCCCGAGACGCCCT
Phenotype
MIM: 608324
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SMIM3 PubMed Links

Gene Details

Gene
SMIM3
Gene Name
small integral membrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032947.4 657 UTR 5 NP_116565.3

View Full Product Details