Product Details
- SNP ID
-
rs2916430
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:4292524 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATTCCATCCTCGCGACAATCTGTT[G/T]CACATGTTAGGCACACAGTATCTGT
- Phenotype
-
MIM: 616238
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LYAR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs570917950] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LYAR
- Gene Name
- Ly1 antibody reactive
There are no transcripts associated with this gene.
- Gene
- ZBTB49
- Gene Name
- zinc finger and BTB domain containing 49
View Full Product Details