Product Details

SNP ID
rs11131190
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:9817514 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CCCAGGCAAATATATGGTGATAGAT[A/G]TGAGCAACTCAGACCTTGCAAGCGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARPC4-TTLL3 PubMed Links

Gene Details

Gene
ARPC4-TTLL3
Gene Name
ARPC4-TTLL3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198793.1 Intron NP_001185722.1
Gene
TTLL3
Gene Name
tubulin tyrosine ligase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025930.3 Intron NP_001021100.3

View Full Product Details