Product Details

SNP ID
rs35792778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2597072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCCTGGTGACAATCTGAAGAGC[A/G]CATCCCCACTGCCTGGTGTGGGTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM193A PubMed Links

Gene Details

Gene
FAM193A
Gene Name
family with sequence similarity 193 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256666.1 Intron NP_001243595.1
NM_001256667.1 Intron NP_001243596.1
NM_001256668.1 Intron NP_001243597.1
NM_003704.3 Intron NP_003695.3
XM_006713930.2 Intron XP_006713993.2
XM_006713932.2 Intron XP_006713995.2
XM_011513590.1 Intron XP_011511892.1
XM_011513593.2 Intron XP_011511895.1
XM_017008788.1 Intron XP_016864277.1

View Full Product Details