Product Details

SNP ID
rs71461328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57559160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAGGGTTGTTCTAATCATCTGCT[A/G]TTACAAACAGTTGAATGACCTTGTA
Phenotype
MIM: 602821
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF5A PubMed Links

Gene Details

Gene
KIF5A
Gene Name
kinesin family member 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004984.2 Intron NP_004975.2

View Full Product Details