Product Details

SNP ID
rs4786275
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:1530024 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ACTTCTCTAACATCTGGGTGGTGGC[A/G]GCACTGGGGCCTCAGTGCGTGAGGC
Phenotype
MIM: 614620 MIM: 611002
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFT140 PubMed Links

Gene Details

Gene
IFT140
Gene Name
intraflagellar transport 140
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014714.3 120 Intron NP_055529.2
XM_005255725.4 120 Intron XP_005255782.1
XM_005255726.3 120 Intron XP_005255783.1
XM_006720990.3 120 Intron XP_006721053.1
XM_006720991.3 120 Intron XP_006721054.1
XM_006720992.3 120 UTR 5 XP_006721055.1
XM_011522766.2 120 Intron XP_011521068.1
XM_011522767.2 120 Intron XP_011521069.1
XM_011522769.2 120 Intron XP_011521071.1
XM_011522771.2 120 Intron XP_011521073.1
XM_011522772.2 120 Intron XP_011521074.1
XM_017023910.1 120 Intron XP_016879399.1
XM_017023911.1 120 Intron XP_016879400.1
Gene
LOC105371047
Gene Name
uncharacterized LOC105371047
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017023923.1 120 Intron XP_016879412.1
Gene
TMEM204
Gene Name
transmembrane protein 204
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256541.1 120 Intron NP_001243470.1
NM_024600.5 120 Intron NP_078876.2

View Full Product Details