Product Details

SNP ID
rs10876478
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:53665955 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AACACCGCTCTTCTAGAAGACAAGA[C/G]TAAGAGGGACAGATTATGTTCTCTC
Phenotype
MIM: 603193
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP5G2 PubMed Links

Gene Details

Gene
ATP5G2
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C2 (subunit 9)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002031.2 Intron NP_001002031.1
NM_005176.5 Intron NP_005167.2
XM_017019460.1 Intron XP_016874949.1
XM_017019461.1 Intron XP_016874950.1
XM_017019462.1 Intron XP_016874951.1

View Full Product Details