Product Details

SNP ID
rs7669853
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:37250008 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCCCCACACAAAAGGCAGACATA[C/T]CTGCATTAAGAAATTCACGTGAAAC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MIR4801 PubMed Links

Gene Details

Gene
MIR4801
Gene Name
microRNA 4801
There are no transcripts associated with this gene.

Gene
NWD2
Gene Name
NACHT and WD repeat domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144990.1 Intron NP_001138462.1

View Full Product Details