Product Details

SNP ID
rs13273801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:56301262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAATGGGCTGTCTTTATTTTTGG[A/G]AAAAAAAAGAAAAAGAAAAAACCAA
Phenotype
MIM: 608989
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDR16C5 PubMed Links

Gene Details

Gene
SDR16C5
Gene Name
short chain dehydrogenase/reductase family 16C, member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318049.1 2027 UTR 3 NP_001304978.1
NM_001318050.1 2027 UTR 3 NP_001304979.1
NM_138969.3 2027 UTR 3 NP_620419.2
XM_011517479.1 2027 UTR 3 XP_011515781.1

View Full Product Details