Product Details

SNP ID
rs35407225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120300802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTAGGTGGTCTTCAAGGTCCCTG[A/C]TGGCCGCCCACCTTTATGTCCAAAG
Phenotype
MIM: 604482
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RNU4-1 PubMed Links

Gene Details

Gene
RNU4-1
Gene Name
RNA, U4 small nuclear 1
There are no transcripts associated with this gene.

Gene
RNU4-2
Gene Name
RNA, U4 small nuclear 2
There are no transcripts associated with this gene.

Gene
SIRT4
Gene Name
sirtuin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012240.2 Intron NP_036372.1
XM_005253865.4 Intron XP_005253922.1
XM_006719308.2 Intron XP_006719371.1
XM_006719309.3 Intron XP_006719372.1

View Full Product Details