Product Details

SNP ID
rs8032727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73684995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGGTTAGGCGTGTCCACTCTGC[A/G]GGGCCCAGCAGGGCGAAGGTGTGGG
Phenotype
MIM: 605715
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD276 PubMed Links

Gene Details

Gene
CD276
Gene Name
CD276 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024736.1 Intron NP_001019907.1
NM_025240.2 Intron NP_079516.1
XM_005254700.4 Intron XP_005254757.1
XM_011522095.2 Intron XP_011520397.1
XM_011522096.2 Intron XP_011520398.1
XM_017022638.1 Intron XP_016878127.1

View Full Product Details