Product Details

SNP ID
rs1877971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:123203028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACATTCGGGTATTTCCTATGTT[G/T]AAAACCATCACATCTGTTTAGTGCC
Phenotype
MIM: 612442
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SEC22A PubMed Links

Gene Details

Gene
SEC22A
Gene Name
SEC22 homolog A, vesicle trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012430.4 Intron NP_036562.2
XM_011512673.2 Intron XP_011510975.1
XM_011512675.2 Intron XP_011510977.1
XM_011512676.2 Intron XP_011510978.1

View Full Product Details