Product Details

SNP ID
rs9151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:163324154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGCACATTAACATGACACAACTA[C/T]TCAGTAATCTTTACAGTGCATTATT
Phenotype
MIM: 162641
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NPY1R PubMed Links

Gene Details

Gene
NPY1R
Gene Name
neuropeptide Y receptor Y1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000909.5 4204 UTR 3 NP_000900.1
XM_005263031.3 4204 UTR 3 XP_005263088.1
XM_011532010.2 4204 UTR 3 XP_011530312.1

View Full Product Details