Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001300816.2 | Intron | NP_001287745.1 | |||
NM_001300817.2 | Intron | NP_001287746.1 | |||
NM_001300818.2 | Intron | NP_001287747.1 | |||
NM_001300819.2 | Intron | NP_001287748.1 | |||
NM_001300822.2 | Intron | NP_001287751.1 | |||
NM_001308127.1 | Intron | NP_001295056.1 | |||
NM_001308128.1 | Intron | NP_001295057.1 | |||
NM_001308129.1 | Intron | NP_001295058.1 | |||
NM_001308130.1 | Intron | NP_001295059.1 | |||
NM_017665.3 | Intron | NP_060135.1 | |||
XM_011543502.1 | Intron | XP_011541804.1 | |||
XM_011543503.2 | Intron | XP_011541805.1 |