Product Details
- SNP ID
-
rs1804142
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:150114107 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGTGCCCAGAGGGGTTGAGCTTC[A/C]TGCACCCCCTTCCCAACCCTCCCCG
- Phenotype
-
MIM: 164770
MIM: 173410
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CSF1R
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143547069] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CSF1R
- Gene Name
- colony stimulating factor 1 receptor
- Gene
- PDGFRB
- Gene Name
- platelet derived growth factor receptor beta
View Full Product Details