Product Details

SNP ID
rs1129647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:161854239 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATTTTGGACAGACTCCTAGATGG[C/T]TATGACAATCGCCTGAGACCAGGAT
Phenotype
MIM: 137160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GABRA1 PubMed Links

Gene Details

Gene
GABRA1
Gene Name
gamma-aminobutyric acid type A receptor alpha1 subunit
There are no transcripts associated with this gene.

View Full Product Details