Product Details

SNP ID
rs2032839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160192205 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGTCCTCTTTTTCTAAATTAAAC[A/T]TTTTAGAGATGGGGTCTTGCTATGT
Phenotype
MIM: 600422
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FABP6 PubMed Links

Gene Details

Gene
FABP6
Gene Name
fatty acid binding protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040442.1 Intron NP_001035532.1
NM_001130958.1 Intron NP_001124430.1
NM_001445.2 Intron NP_001436.1

View Full Product Details