Product Details

SNP ID
rs439418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:117265911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGTTTGCGGCGGCATGGCCTGT[A/G]CGCCAAGGTCTGCTGTGCCTCCGCG
Phenotype
MIM: 609979
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VGLL2 PubMed Links

Gene Details

Gene
VGLL2
Gene Name
vestigial like family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153453.1 Intron NP_703154.1
NM_182645.3 Intron NP_872586.1
XM_005266883.2 Intron XP_005266940.1

View Full Product Details