Product Details

SNP ID
rs1978602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176370486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACTTGACATGGAGAGAGACTGT[T/A]TGTCTTCGACCTCAGATTCCCCAAC
Phenotype
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
ARL10 PubMed Links

Gene Details

Gene
ARL10
Gene Name
ADP ribosylation factor like GTPase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317948.1 Intron NP_001304877.1
NM_173664.5 Intron NP_775935.1
XM_011534529.2 Intron XP_011532831.1
XM_011534530.2 Intron XP_011532832.1
XM_011534531.2 Intron XP_011532833.1
XM_017009372.1 Intron XP_016864861.1
Gene
KIAA1191
Gene Name
KIAA1191
There are no transcripts associated with this gene.

Gene
MIR1271
Gene Name
microRNA 1271
There are no transcripts associated with this gene.

View Full Product Details