Product Details

SNP ID
rs13713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:172044475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGAAAAACCAAGAATTCCAGCC[C/T]GGATGAGCCCACGGGGACTTCATCA
Phenotype
MIM: 603919
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STK10 PubMed Links

Gene Details

Gene
STK10
Gene Name
serine/threonine kinase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005990.3 3614 UTR 3 NP_005981.3
XM_017009788.1 3614 UTR 3 XP_016865277.1

View Full Product Details