Product Details
- SNP ID
-
rs1830519
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:160711798 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTGGGACTGAAGTAAGCATATCA[A/G]GTTAGAACTCTCATCACATGTTCGA
- Phenotype
-
MIM: 173350
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLG
PubMed Links
Gene Details
- Gene
- PLG
- Gene Name
- plasminogen
View Full Product Details