Product Details

SNP ID
rs10635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:98041748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAAAACAAGCAAATTTTATTAAA[A/G]GAAAATTTTGCAGGTTTAAGGTTTG
Phenotype
MIM: 602108 MIM: 180467
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MATN2 PubMed Links

Gene Details

Gene
MATN2
Gene Name
matrilin 2
There are no transcripts associated with this gene.

Gene
RPL30
Gene Name
ribosomal protein L30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000989.3 Intron NP_000980.1
Gene
SNORA72
Gene Name
small nucleolar RNA, H/ACA box 72
There are no transcripts associated with this gene.

View Full Product Details