Product Details

SNP ID
rs7516736
Assay Type
Functionally tested
NCBI dbSNP Submissions
51
Location
Chr.1:156825994 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAAAGGAAGTGGGGATTAGAACC[A/G]GATGGTCTGGTTTCAGATTTTGGGA
Phenotype
MIM: 191315 MIM: 604514
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NTRK1 PubMed Links

Gene Details

Gene
NTRK1
Gene Name
neurotrophic receptor tyrosine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007792.1 Intron NP_001007793.1
NM_001012331.1 Intron NP_001012331.1
NM_002529.3 Intron NP_002520.2
Gene
SH2D2A
Gene Name
SH2 domain containing 2A
There are no transcripts associated with this gene.

View Full Product Details