Product Details

SNP ID
rs1898466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:84185557 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGAATCTGCTGGAAATTTTTCTGTC[C/T]AAATAAATTTCCTGAAGGCATACAT
Phenotype
MIM: 614648
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RALYL PubMed Links

Gene Details

Gene
RALYL
Gene Name
RALY RNA binding protein-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100391.1 Intron NP_001093861.1
NM_001100392.1 Intron NP_001093862.1
NM_001100393.1 Intron NP_001093863.1
NM_001287243.1 Intron NP_001274172.1
NM_001287244.1 Intron NP_001274173.1
NM_173848.5 Intron NP_776247.3
XM_011517462.2 Intron XP_011515764.1
XM_017013058.1 Intron XP_016868547.1
XM_017013059.1 Intron XP_016868548.1
XM_017013060.1 Intron XP_016868549.1
XM_017013061.1 Intron XP_016868550.1
XM_017013062.1 Intron XP_016868551.1
XM_017013063.1 Intron XP_016868552.1
XM_017013064.1 Intron XP_016868553.1
XM_017013065.1 Intron XP_016868554.1
XM_017013066.1 Intron XP_016868555.1
XM_017013067.1 Intron XP_016868556.1
XM_017013068.1 Intron XP_016868557.1
XM_017013069.1 Intron XP_016868558.1
XM_017013070.1 Intron XP_016868559.1
XM_017013071.1 Intron XP_016868560.1
XM_017013072.1 Intron XP_016868561.1
XM_017013073.1 Intron XP_016868562.1
XM_017013074.1 Intron XP_016868563.1
XM_017013075.1 Intron XP_016868564.1
XM_017013076.1 Intron XP_016868565.1
XM_017013077.1 Intron XP_016868566.1
XM_017013078.1 Intron XP_016868567.1
XM_017013079.1 Intron XP_016868568.1
XM_017013080.1 Intron XP_016868569.1
XM_017013081.1 Intron XP_016868570.1
XM_017013082.1 Intron XP_016868571.1
XM_017013083.1 Intron XP_016868572.1
XM_017013084.1 Intron XP_016868573.1
XM_017013085.1 Intron XP_016868574.1
XM_017013086.1 Intron XP_016868575.1
XM_017013087.1 Intron XP_016868576.1

View Full Product Details