Product Details
- SNP ID
-
rs1820851
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:22095238 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGGAAGAACAAGGCCATGTCCAC[A/G]TGCACCACTCGTGTCATTATTATAC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101927079
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74002754] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101927079
- Gene Name
- uncharacterized LOC101927079
There are no transcripts associated with this gene.
- Gene
- OR4N4
- Gene Name
- olfactory receptor family 4 subfamily N member 4
View Full Product Details