Product Details

SNP ID
rs2300295
Assay Type
Validated
NCBI dbSNP Submissions
55
Location
Chr.1:168577513 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
TTCAGTAGGTATTTCTTAAGGATTA[A/C]TTTTTCCTATGACTGGAGTGAATCT
Phenotype
MIM: 600250
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
XCL1 PubMed Links

Gene Details

Gene
XCL1
Gene Name
X-C motif chemokine ligand 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002995.2 Intron NP_002986.1
XM_011509865.2 Intron XP_011508167.1

View Full Product Details