Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001318533.1 | Intron | NP_001305462.1 | |||
NM_017868.3 | Intron | NP_060338.3 | |||
XM_005271604.3 | Intron | XP_005271661.1 | |||
XM_011542890.2 | Intron | XP_011541192.1 | |||
XM_017017943.1 | Intron | XP_016873432.1 | |||
XM_017017944.1 | Intron | XP_016873433.1 | |||
XM_017017945.1 | Intron | XP_016873434.1 | |||
XM_017017946.1 | Intron | XP_016873435.1 | |||
XM_017017947.1 | Intron | XP_016873436.1 | |||
XM_017017948.1 | Intron | XP_016873437.1 | |||
XM_017017949.1 | Intron | XP_016873438.1 | |||
XM_017017950.1 | Intron | XP_016873439.1 |