Product Details

SNP ID
rs7170
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:36393562 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTCACCTACCCCAAGGCCACATA[C/T]AGGATTCTGGGTCCCAGAGTGAGTC
Phenotype
MIM: 609437
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LSM10 PubMed Links

Gene Details

Gene
LSM10
Gene Name
LSM10, U7 small nuclear RNA associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032881.2 785 UTR 3 NP_116270.1
Gene
STK40
Gene Name
serine/threonine kinase 40
There are no transcripts associated with this gene.

View Full Product Details