Product Details

SNP ID
rs980
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:37862320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAGGAGGAGAGAGAGGCTCAGA[G/T]TGGGTTGCAGAGGAACTGGTGAATA
Phenotype
MIM: 147264 MIM: 600172
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
INPP5B PubMed Links

Gene Details

Gene
INPP5B
Gene Name
inositol polyphosphate-5-phosphatase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297434.1 2571 Missense Mutation NP_001284363.1
NM_005540.2 2571 Missense Mutation NP_005531.2
XM_006710621.3 2571 Intron XP_006710684.2
XM_011541392.2 2571 Missense Mutation XP_011539694.1
XM_011541393.2 2571 Missense Mutation XP_011539695.1
XM_017001206.1 2571 Intron XP_016856695.1
XM_017001207.1 2571 Missense Mutation XP_016856696.1
XM_017001208.1 2571 Intron XP_016856697.1
XM_017001209.1 2571 Missense Mutation XP_016856698.1
Gene
MTF1
Gene Name
metal regulatory transcription factor 1
There are no transcripts associated with this gene.

View Full Product Details