Product Details

SNP ID
rs1983478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:130954899 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTTGAGCTGGGTCATATGAAACA[C/T]TGTGATTAAGGAAATATGATCTTCA
Phenotype
MIM: 613639
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADGRD1 PubMed Links

Gene Details

Gene
ADGRD1
Gene Name
adhesion G protein-coupled receptor D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005253566.2 Intron XP_005253623.1
XM_011538203.2 Intron XP_011536505.1
XM_011538204.2 Intron XP_011536506.1
XM_011538205.2 Intron XP_011536507.1
XM_011538206.2 Intron XP_011536508.1
XM_011538207.2 Intron XP_011536509.1
XM_011538208.2 Intron XP_011536510.1
XM_011538209.2 Intron XP_011536511.1
XM_011538210.2 Intron XP_011536512.1
XM_011538211.2 Intron XP_011536513.1
XM_011538212.2 Intron XP_011536514.1

View Full Product Details