Product Details

SNP ID
rs11301777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:105370667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATGCATTCTCCTTATTTTCTGA[A/G]TTAGAAAATATTTCATCTGAAATTT
Phenotype
MIM: 609988
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPA2 PubMed Links

Gene Details

Gene
PPA2
Gene Name
pyrophosphatase (inorganic) 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006903.4 Intron NP_008834.3
NM_176866.2 Intron NP_789842.2
NM_176867.3 Intron NP_789843.2
NM_176869.2 Intron NP_789845.1

View Full Product Details