Product Details

SNP ID
rs1823251
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:44303615 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AACTCTTATGTACTCTTTTCTATAC[C/G]TGCTCTAACAAAATGAATTTTGATA
Phenotype
MIM: 602115
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FGF10 PubMed Links

Gene Details

Gene
FGF10
Gene Name
fibroblast growth factor 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004465.1 3014 Intron NP_004456.1
XM_005248264.3 3014 UTR 3 XP_005248321.1

View Full Product Details