Product Details

SNP ID
rs13810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11548954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATCTGGCACAGAGAAACAAGGG[A/G]AGACAGGGCAGTGATAAGATCCAGC
Phenotype
MIM: 603795
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LITAF PubMed Links

Gene Details

Gene
LITAF
Gene Name
lipopolysaccharide induced TNF factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136472.1 1317 UTR 3 NP_001129944.1
NM_001136473.1 1317 UTR 3 NP_001129945.1
NM_004862.3 1317 UTR 3 NP_004853.2
XM_006720982.2 1317 UTR 3 XP_006721045.1
XM_006720983.3 1317 UTR 3 XP_006721046.1
XM_006720984.3 1317 UTR 3 XP_006721047.1
XM_006720985.3 1317 UTR 3 XP_006721048.1
XM_011522754.2 1317 UTR 3 XP_011521056.1
XM_017023896.1 1317 UTR 3 XP_016879385.1

View Full Product Details