Product Details

SNP ID
rs11053547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10012166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATAAATAGCATGAATAATAGAT[A/G]AGTTATTGTTAACAGGTAAAAGATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLEC12B PubMed Links

Gene Details

Gene
CLEC12B
Gene Name
C-type lectin domain family 12 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129998.2 Intron NP_001123470.1
NM_001319241.1 Intron NP_001306170.1
NM_001319242.1 Intron NP_001306171.1
NM_205852.3 Intron NP_995324.2
XM_011520658.2 Intron XP_011518960.1
XM_011520663.2 Intron XP_011518965.1
XM_017019295.1 Intron XP_016874784.1
Gene
LOC102724020
Gene Name
uncharacterized LOC102724020
There are no transcripts associated with this gene.

View Full Product Details