Product Details

SNP ID
rs876856
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:19861207 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCAGACCTTGTTTCTGAAAGAAGCA[T/G]GTAGCTATGTTTGGAAATTAAATGT
Phenotype
MIM: 605948
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
GPRC5B PubMed Links

Gene Details

Gene
GPRC5B
Gene Name
G protein-coupled receptor class C group 5 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304771.1 Intron NP_001291700.1
NM_016235.2 Intron NP_057319.1
XM_006721051.2 Intron XP_006721114.1
XM_006721052.2 Intron XP_006721115.1
Gene
IQCK
Gene Name
IQ motif containing K
There are no transcripts associated with this gene.

View Full Product Details