Product Details

SNP ID
rs2901647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:64190963 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGAAGCTTAGGCCTTGTTAGCCT[C/G]CTGCCCACAGTAAACACCTGGAGAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00309 PubMed Links

Gene Details

Gene
LINC00309
Gene Name
long intergenic non-protein coding RNA 309
There are no transcripts associated with this gene.

View Full Product Details