Product Details

SNP ID
rs2581456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:41997376 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTAGACTATATTACAAATTGTGAT[A/G]ATGATGTTAATAAGACCTTGATGAA
Phenotype
MIM: 604604
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC30A9 PubMed Links

Gene Details

Gene
SLC30A9
Gene Name
solute carrier family 30 member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006345.3 Intron NP_006336.3
XM_017007654.1 Intron XP_016863143.1

View Full Product Details