Product Details

SNP ID
hCV11366393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:71199340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGCTGTCAAAGTGCCGAGCGCTG[G/T]TACAGGTACTCCAGTTCCAAGGCAT
Phenotype
MIM: 601653
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EYA1 PubMed Links

Gene Details

Gene
EYA1
Gene Name
EYA transcriptional coactivator and phosphatase 1
There are no transcripts associated with this gene.

View Full Product Details