Product Details

SNP ID
rs3395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:67422176 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATCATTACTAGCCTAGATCCTAA[C/T]TGAGGACATGAGATTTATTGAAGGG
Phenotype
MIM: 609562
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPA6 PubMed Links

Gene Details

Gene
CPA6
Gene Name
carboxypeptidase A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020361.4 1898 UTR 3 NP_065094.3
XM_011517570.2 1898 UTR 3 XP_011515872.1
XM_017013646.1 1898 UTR 3 XP_016869135.1
XM_017013647.1 1898 Intron XP_016869136.1
Gene
LOC102724708
Gene Name
uncharacterized LOC102724708
There are no transcripts associated with this gene.

View Full Product Details