Product Details
- SNP ID
-
rs3104233
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:55108129 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTATGTTTTTCTTCCAACAGCTGA[C/T]GGAAAAAGCAAAAATTAACAAATAT
- Phenotype
-
MIM: 140050
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDC20B
PubMed Links
Gene Details
- Gene
- CDC20B
- Gene Name
- cell division cycle 20B
There are no transcripts associated with this gene.
- Gene
- GZMA
- Gene Name
- granzyme A
There are no transcripts associated with this gene.
View Full Product Details