Product Details
- SNP ID
-
rs3255
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:5393218 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TACCAGAAAGGAAAAGCATAAACTT[C/T]AGGATTTCATTGTCTCTTGGTAAAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TUBAL3
PubMed Links
Gene Details
- Gene
- TUBAL3
- Gene Name
- tubulin alpha like 3
View Full Product Details