Product Details

SNP ID
rs10925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:15211996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACGTTACACATGATAACTAGAGA[G/T]CACACTGTGTTGAAACGAGGATGCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM171A1 PubMed Links

Gene Details

Gene
FAM171A1
Gene Name
family with sequence similarity 171 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010924.1 3520 UTR 3 NP_001010924.1
XM_011519378.2 3520 UTR 3 XP_011517680.1
XM_017015904.1 3520 UTR 3 XP_016871393.1
XM_017015905.1 3520 UTR 3 XP_016871394.1

View Full Product Details