Product Details

SNP ID
rs2050354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:7561914 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCCCAAGCGCTCCCGGTATCCGTC[C/G]GCTGAAGGGTGACGTTCATTGACTC
Phenotype
MIM: 609783
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ITIH5 PubMed Links

Gene Details

Gene
ITIH5
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001851.2 4104 Intron NP_001001851.1
NM_030569.6 4104 UTR 3 NP_085046.5
NM_032817.5 4104 UTR 3 NP_116206.4
XM_011519713.2 4104 UTR 3 XP_011518015.1
XM_011519714.2 4104 Intron XP_011518016.1

View Full Product Details