Product Details

SNP ID
rs566620411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74892706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCAGGCTCCGCAGCACTACCCC[A/G]ATGCCAACCACAAGCCAGAGATGGC
Phenotype
MIM: 154550
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM219B PubMed Links

Gene Details

Gene
FAM219B
Gene Name
family with sequence similarity 219 member B
There are no transcripts associated with this gene.

Gene
MPI
Gene Name
mannose phosphate isomerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289155.1 326 Missense Mutation AAT,GAT N131D NP_001276084.1
NM_001289156.1 326 Missense Mutation AAT,GAT N81D NP_001276085.1
NM_001289157.1 326 Missense Mutation AAT,GAT N131D NP_001276086.1
NM_002435.2 326 Missense Mutation AAT,GAT N131D NP_002426.1
XM_011521592.1 326 Missense Mutation AAT,GAT N127D XP_011519894.1
XM_011521593.2 326 Missense Mutation AAT,GAT N111D XP_011519895.1
XM_017022207.1 326 Missense Mutation AAT,GAT N127D XP_016877696.1
XM_017022208.1 326 Missense Mutation AAT,GAT N111D XP_016877697.1
XM_017022209.1 326 Missense Mutation AAT,GAT N81D XP_016877698.1

View Full Product Details