Product Details

SNP ID
rs872029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81934710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCCTTGGAAGTCCCATCTTCAC[G/A]CCCCCATCAGCCAGGGCATCCAGGG
Phenotype
MIM: 602020 MIM: 179035
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
MAFG PubMed Links

Gene Details

Gene
MAFG
Gene Name
MAF bZIP transcription factor G
There are no transcripts associated with this gene.

Gene
MAFG-AS1
Gene Name
MAFG antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
MYADML2
Gene Name
myeloid associated differentiation marker like 2
There are no transcripts associated with this gene.

Gene
PYCR1
Gene Name
pyrroline-5-carboxylate reductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282279.1 648 Intron NP_001269208.1
NM_001282280.1 648 Silent Mutation GGC,GGT G192G NP_001269209.1
NM_001282281.1 648 Silent Mutation GGC,GGT G219G NP_001269210.1
NM_006907.3 648 Silent Mutation GGC,GGT G192G NP_008838.2
NM_153824.2 648 Silent Mutation GGC,GGT G192G NP_722546.1
XM_005256381.2 648 Silent Mutation GGC,GGT G192G XP_005256438.1
XM_011523583.2 648 Silent Mutation GGC,GGT G192G XP_011521885.1
XM_011523584.2 648 Silent Mutation GGC,GGT G192G XP_011521886.1
XM_011523585.2 648 Silent Mutation GGC,GGT G219G XP_011521887.1
XM_017024909.1 648 Silent Mutation GGC,GGT G192G XP_016880398.1

View Full Product Details