Product Details

SNP ID
rs5384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81812864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGGGCGTCCACGAAGTGGTCTT[C/T]GCCTTCGTGACGGACGAGCACGCCC
Phenotype
MIM: 138033 MIM: 616514
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCGR PubMed Links

Gene Details

Gene
GCGR
Gene Name
glucagon receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000160.4 1390 Silent Mutation TTC,TTT F365F NP_000151.1
XM_006722277.1 1390 Silent Mutation TTC,TTT F365F XP_006722340.1
XM_011523539.1 1390 Silent Mutation TTC,TTT F332F XP_011521841.1
XM_017024446.1 1390 Silent Mutation TTC,TTT F363F XP_016879935.1
XM_017024447.1 1390 Silent Mutation TTC,TTT F243F XP_016879936.1
Gene
MCRIP1
Gene Name
MAPK regulated corepressor interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details