Product Details

SNP ID
rs8360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63976358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCCTCACACTGTGACCTGACCA[C/G]CCCCACCGGCCCATCCTGGTCATGT
Phenotype
MIM: 123870 MIM: 608337
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COX8A PubMed Links

Gene Details

Gene
COX8A
Gene Name
cytochrome c oxidase subunit 8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004074.2 322 UTR 3 NP_004065.1
Gene
OTUB1
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 1
There are no transcripts associated with this gene.

View Full Product Details