Product Details

SNP ID
rs630437
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65792670 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGAATGAGGTGGCTCATTAAAGG[C/T]CCCCGGACAGACGGACGCCAGTGCC
Phenotype
MIM: 602313
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OVOL1 PubMed Links

Gene Details

Gene
OVOL1
Gene Name
ovo like transcriptional repressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004561.3 Intron NP_004552.2
XM_005274018.3 Intron XP_005274075.1
XM_011545067.2 Intron XP_011543369.1
XM_011545068.2 Intron XP_011543370.1
XM_017017837.1 Intron XP_016873326.1
XM_017017838.1 Intron XP_016873327.1
Gene
OVOL1-AS1
Gene Name
OVOL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details