Product Details

SNP ID
hCV11396301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100306884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCCAGTATCTTATAGAATATCC[A/G]TAATATAGATTCATAAATAAACCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPDYE3 PubMed Links

Gene Details

Gene
SPDYE3
Gene Name
speedy/RINGO cell cycle regulator family member E3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004351.4 Intron NP_001004351.3

View Full Product Details