Product Details

SNP ID
rs2063979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35976530 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAAGGGAGTTTTATTTATTGCATG[C/T]TTGGGTTTTGGACTAAGAGTTTACC
Phenotype
MIM: 601392 MIM: 601394
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL14 PubMed Links

Gene Details

Gene
CCL14
Gene Name
C-C motif chemokine ligand 14
There are no transcripts associated with this gene.

Gene
CCL15-CCL14
Gene Name
CCL15-CCL14 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
CCL16
Gene Name
C-C motif chemokine ligand 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004590.3 1475 UTR 3 NP_004581.1
XM_005258020.4 1475 UTR 3 XP_005258077.1
Gene
LOC101927339
Gene Name
uncharacterized LOC101927339
There are no transcripts associated with this gene.

View Full Product Details